Learn More
Invitrogen™ Human CDK3/cyclin E1, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human CDK3 and Cyclin E1 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for the subunits CDK3 and Cyclin E1 are NP_001249.1 and NP_001229.1.
Specifications
Specifications
| Accession Number | P24864, Q00526 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 25% glycerol, 0.1mM EDTA, 0.1mM PMSF, 150mM NaCl, 0.25mM DTT, 0.1mM EGTA and no preservative; pH 7.5 |
| Gene ID (Entrez) | 1018, 898 |
| Molecular Weight (g/mol) | 61.3-73.4 kDa |
| Name | Human CDK3/cyclin E1, GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.