Learn More
Medchemexpress LLC Cdk4/6-in-15 | 00-00-0 | 99.3% | 442.56 g/mol | C21H27FN8S | 5 MG
SDP
Catalog No. 5000221355
Encompass_Preferred
CDK4/6-IN-15 is an orally active, selective inhibitor of cyclin-dependent kinases CDK4 and CDK6 used in cellular and biochemical research to probe cell-cycle regulation and anticancer activity. It arrests cells in G1, suppresses Rb phosphorylation, and has demonstrated antiproliferative and pro-apoptotic effects in multiple cell lines.
  • selective inhibitor of CDK4 and CDK6 with potent activity
  • arrests the cell cycle at G1 and reduces Rb phosphorylation
  • demonstrated antiproliferative and pro-apoptotic effects in cell assays
  • soluble in DMSO with stock preparation guidance for common concentrations
  • stable when protected from light; storage recommendations provided
Catalog No. 50-002-21355 Supplier Medchemexpress LLC Supplier No. HY142076A5MG
CDK4/6-IN-15 is an orally active, selective inhibitor of cyclin-dependent kinases CDK4 and CDK6 used in cellular and biochemical research to probe cell-cycle regulation and anticancer activity. It arrests cells in G1, suppresses Rb phosphorylation, and has demonstrated antiproliferative and pro-apoptotic effects in multiple cell lines.
  • selective inhibitor of CDK4 and CDK6 with potent activity
  • arrests the cell cycle at G1 and reduces Rb phosphorylation
  • demonstrated antiproliferative and pro-apoptotic effects in cell assays
  • soluble in DMSO with stock preparation guidance for common concentrations
  • stable when protected from light; storage recommendations provided