Learn More
Medchemexpress LLC Cdk4/6-in-15 hydrochloride | 00-00-0 | 99.3% | C21H27FN8S | 10MG
SDP
Catalog No. 5000261060
Encompass_Preferred
CDK4/6-IN-15 hydrochloride is an orally active, selective inhibitor of cyclin-dependent kinases 4 and 6 that arrests the cell cycle at G1 and exhibits antiproliferative activity in cancer cell models. The hydrochloride salt is provided as a high-purity research reagent intended for in vitro cell-cycle and oncology studies.
  • Selective CDK4/6 inhibition for targeted cell-cycle studies.
  • Arrests cells in G1 phase to assess proliferation effects.
  • Hydrochloride salt form for improved solubility in aqueous media.
  • High purity (≈99.3%) suitable for research applications.
  • Supplied in small-scale packaging for pilot experiments.
Catalog No. 50-002-61060 Supplier Medchemexpress LLC Supplier No. HY142076A10MG
Add to Cart
Add to Cart
CDK4/6-IN-15 hydrochloride is an orally active, selective inhibitor of cyclin-dependent kinases 4 and 6 that arrests the cell cycle at G1 and exhibits antiproliferative activity in cancer cell models. The hydrochloride salt is provided as a high-purity research reagent intended for in vitro cell-cycle and oncology studies.
  • Selective CDK4/6 inhibition for targeted cell-cycle studies.
  • Arrests cells in G1 phase to assess proliferation effects.
  • Hydrochloride salt form for improved solubility in aqueous media.
  • High purity (≈99.3%) suitable for research applications.
  • Supplied in small-scale packaging for pilot experiments.