Learn More
Medchemexpress LLC CDK4/6-IN-15 (hydrochloride) | 2078047-99-9 | MFCD09904297 | 99.3% | C21H28ClFN8S | 100 MG
SDP
Catalog No. 5000353300
Encompass_Preferred
CDK4/6-IN-15 (hydrochloride) is an orally active and selective CDK4/6 inhibitor that potently inhibits cancer cell growth. It arrests the cell cycle at the G1 phase and suppresses retinoblastoma tumour suppressor protein (Rb) phosphorylation at S780 and E2 factor (E2F)-regulated gene expression.
  • Potently inhibits cancer cell growth
  • Arrests the cell cycle at G1 phase
  • Suppresses retinoblastoma tumour suppressor protein (Rb) phosphorylation at S780
  • Suppresses E2 factor (E2F)-regulated gene expression
Catalog No. 50-003-53300 Supplier Medchemexpress LLC Supplier No. HY142076A100MG
CDK4/6-IN-15 (hydrochloride) is an orally active and selective CDK4/6 inhibitor that potently inhibits cancer cell growth. It arrests the cell cycle at the G1 phase and suppresses retinoblastoma tumour suppressor protein (Rb) phosphorylation at S780 and E2 factor (E2F)-regulated gene expression.
  • Potently inhibits cancer cell growth
  • Arrests the cell cycle at G1 phase
  • Suppresses retinoblastoma tumour suppressor protein (Rb) phosphorylation at S780
  • Suppresses E2 factor (E2F)-regulated gene expression