Learn More
Medchemexpress LLC HY-114339 10mg Medchemexpress, CDK4/6-IN-2 CAS:1800506-48-2 Purity:>98%
SDP
Catalog No. 501969331
Encompass_Preferred
Medchemexpress, HY-114339 10mg CDK4/6-IN-2 CAS:1800506-48-2 CDK4/6-IN-2 is a potent CDK4 and CDK6 inhibitor extracted from patent US20180000819A1, Compound 1, has IC50s of 2.7 and 16 nM for CDK4 and CDK6, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9331 Supplier Medchemexpress LLC Supplier No. HY11433910MG
Medchemexpress, HY-114339 10mg CDK4/6-IN-2 CAS:1800506-48-2 CDK4/6-IN-2 is a potent CDK4 and CDK6 inhibitor extracted from patent US20180000819A1, Compound 1, has IC50s of 2.7 and 16 nM for CDK4 and CDK6, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.