Learn More
Medchemexpress LLC HY-129336 5mg Medchemexpress, CDK4/6-IN-4 CAS:2138499-06-4 Purity:>98%
SDP
Catalog No. 501969803
Encompass_Preferred
Medchemexpress, HY-129336 5mg CDK4/6-IN-4 CAS:2138499-06-4 CDK4/6-IN-4, the active metabolite of Abemaciclib, is a selective CDK4/6 inhibitor for the treatment of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9803 Supplier Medchemexpress LLC Supplier No. HY1293365MG
Medchemexpress, HY-129336 5mg CDK4/6-IN-4 CAS:2138499-06-4 CDK4/6-IN-4, the active metabolite of Abemaciclib, is a selective CDK4/6 inhibitor for the treatment of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.