Learn More
Abnova™ CDK4/CEN12 CISH Probe
Catalog No. 89335536 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-P
Catalog No. For Use With (Application)
89-335-536 CISH-P
1 options
Catalog No. 89-335-536 Supplier Abnova™ Supplier No. CG0003
Add to Cart
Add to Cart
CDK4/CEN12 CISH Probe is designed for the qualitative detection of human CDK4 gene as well as chromosome 12 alpha satellites in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH).
The probe is intended to be used in combination with the CISH Implementation Kit 2, which provides necessary reagents for specimen pretreatment and post-hybridization processing.
Hybridization signals of digoxigenin-labeled polynucleotides appear as dark green colored distinct dots (CDK4 gene), and dinitrophenyl-labeled polynucleotides appear as bright red colored distinct dots (CEN 12).
Normal situation: In interphases of normal cells or cells without an amplification involving the CDK4 gene, two green and two red signals appear.
Aberrant situation: In a cell with amplification of the CDK4 gene locus, multiple copies of the green signal or green signal clusters will be observed.
Other signal distribution may be observed in some abnormal samples which might result in a different signal pattern than described above, indicating variant rearrangements.
Unexpected signal patterns should be further investigated.
Specifications
Content And Storage The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use.
For Use With (Application) CISH-P