Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CDK4/cyclin D3, GST Tag Recombinant Protein

Catalog No. A33736
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A33736 1 mg
A33515 10 μg
A33573 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A33736 Supplier Invitrogen™ Supplier No. A33736
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This CDK4/cyclin D3 recombinant human protein (CDK4: full length, cyclin D3: full length) was expressed in insect cells. CDK4/cyclin D3 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for the subunits CKD4 and Cyclin D3 are NP_000066.1 and NP_001751.1.

CDK4, also known as Cyclin-Dependent Kinase 4, is a member of the cyclin-dependent kinase (CDK) family. These kinases are key regulators of cell cycle progression. CDK4 is particularly involved in driving the transition from the G1 phase to the S phase of the cell cycle. Cyclin D3 is a regulatory subunit that binds to CDK4 and activates its kinase activity. The binding of Cyclin D3 to CDK4 is essential for the proper functioning of CDK4, as cyclins regulate the activation and specificity of CDKs. Together, the CDK4/Cyclin D3 complex plays a crucial role in promoting cell cycle progression, specifically by phosphorylating the retinoblastoma protein (Rb), which leads to the release of E2F transcription factors and the subsequent activation of genes required for DNA synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11802, P30281
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.1mM PMSF, 150mM NaCl, 0.1mM EDTA, 0.25mM DTT, 25% glycerol, 10mM glutathione and no preservative; pH 7.5
Gene ID (Entrez) 1019, 896
Molecular Weight (g/mol) 60.0-58.8 kDa
Name Human CDK4/cyclin D3, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Cdk4; Cell division protein kinase 4; CMM3; Crk3; cyclin dependent kinase 4; cyclin-dependent kinase 4; MGC14458; PSK-J3; serine/threonine kinase; Unknown (protein for MGC:133903)
Common Name CDK4/Cyclin D3
Gene Symbol CCND3, CDK4
Kinase Family CDKs and CHKs
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence CDK4: 1-303, Cyclin D3: 1-292
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.