Learn More
Sino Biological Recombinant Human CDK4 Protein (full length, GST Tag)
Catalog No. 5000083937
Encompass_Preferred
Recombinant full-length human CDK4 and CyclinD2 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.
Catalog No. 50-000-83937 Supplier Sino Biological Supplier No. C3118BG50
Recombinant full-length human CDK4 and CyclinD2 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.