Learn More
Sino Biological Recombinant Human CDK4 Protein (full length, GST Tag)
Catalog No. 5000084129
Encompass_Preferred
Recombinant full-length human CDK4 and CyclinD2 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.
Catalog No. 50-000-84129 Supplier Sino Biological Supplier No. C3118BG500
Recombinant full-length human CDK4 and CyclinD2 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.