Learn More
Cayman Chemical Cdk4 InhIbItr 500ug
SDP
Catalog No. 502967295
Encompass_Preferred
A cell-permeable, asymmetrical indolocarbazole that exhibits antiproliferative activity by selectively blocking cyclin D1/Cdk4 (IC50 = 0.8 μM); inhibits HCT116 and NCI-H460 tumor cell growth (IC50s 1 cell cycle arrest
Catalog No. 50-296-7295 Supplier Cayman Chemical Supplier No. 17648500 UG
Add to Cart
Add to Cart
A cell-permeable, asymmetrical indolocarbazole that exhibits antiproliferative activity by selectively blocking cyclin D1/Cdk4 (IC50 = 0.8 μM); inhibits HCT116 and NCI-H460 tumor cell growth (IC50s 1 cell cycle arrest