Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP10035BA20UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His. Host: Baculovirus. Species Reactivity: Human. Protein Expression: A DNA sequence encoding the Homo sapiens (Human) CDK4, was expressed in the hosts and tags indicated. AA Sequence: MATSRYEPVA EIGVGAYGTV YKARDPHSGH FVALKSVRVP NGGGGGGGLP ISTVREVALL RRLEAFEHPN VVRLMDVCAT SRTDREIKRC FVESLSSVET LKPTSWAKSL T. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: CDK4. Uniprot: Q96BE9.
Catalog No. 50-188-5481