Learn More
Cayman Chemical Cdk4/6 Inhibitor IV, Purity: ≥98%, CAS: 359886-84-3, 5 mg
SDP
Catalog No. 502966031
Encompass_Preferred
A cell-permeable triaminopyrimidine that selectively and reversibly blocks Cdk4/cyclin D1 and Cdk6/cyclin D1 activity (IC50s = 1.5 and 5.6 μM, respectively); induces cell cycle arrest in the G1 phase and apoptosis in asynchronous cell lines; suppresses tumor growth in mice bearing human HCT116 colon carcinoma xenografts
Catalog No. 50-296-6031 Supplier Cayman Chemical Supplier No. 179745 MG
A cell-permeable triaminopyrimidine that selectively and reversibly blocks Cdk4/cyclin D1 and Cdk6/cyclin D1 activity (IC50s = 1.5 and 5.6 μM, respectively); induces cell cycle arrest in the G1 phase and apoptosis in asynchronous cell lines; suppresses tumor growth in mice bearing human HCT116 colon carcinoma xenografts