Learn More
Aviva Systems Biology Corp CDK5 Antibody - C-terminal region (ARP38526_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38526_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CDK5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CDK5. Alternate names: PSSALREApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-56688