Learn More
Medchemexpress LLC CDK5-IN-1 | 2639540-19-3 | 98.5% | 496.56 | 50 MG
SDP
Catalog No. 5000350915
Encompass_Preferred
CDK5-IN-1 is a potent CDK5 inhibitor with an IC50 less than 10 nM. It is more than 100-fold more active against CDK5 than CDK2 and is used for kidney diseases research.
  • Potent CDK5 inhibitor with an IC50 less than 10 nM
  • More than 100-fold more active against CDK5 than CDK2
  • Used for kidney diseases research
Catalog No. 50-003-50915 Supplier Medchemexpress LLC Supplier No. HY13972550MG
CDK5-IN-1 is a potent CDK5 inhibitor with an IC50 less than 10 nM. It is more than 100-fold more active against CDK5 than CDK2 and is used for kidney diseases research.
  • Potent CDK5 inhibitor with an IC50 less than 10 nM
  • More than 100-fold more active against CDK5 than CDK2
  • Used for kidney diseases research