Learn More
Medchemexpress LLC Cdk5 inhibitor 20-223 | 865317-30-2 | MFCD32857141 | 99.6% | 305.37 | C19H19N3O | 25MG
SDP
Catalog No. 5000237495
Encompass_Preferred
CDK5 inhibitor 20-223 is a small-molecule kinase inhibitor active against CDK2 and CDK5, with low-nanomolar enzymatic potency and demonstrated cellular activity in colorectal cancer models. It is supplied as a light yellow to yellow solid for laboratory research, provided with high purity and recommended cold storage for long-term stability.
  • Potent CDK2 and CDK5 inhibition with IC50s of ~6.0 nM and ~8.8 nM.
  • Demonstrated cellular activity in colorectal cancer cell lines.
  • High purity suitable for research applications.
  • Light yellow to yellow solid for ease of handling.
  • Molecular formula C19H19N3O and molecular weight 305.37.
  • Recommended cold storage for long-term stability.
Catalog No. 50-002-37495 Supplier Medchemexpress LLC Supplier No. HY12377225MG
CDK5 inhibitor 20-223 is a small-molecule kinase inhibitor active against CDK2 and CDK5, with low-nanomolar enzymatic potency and demonstrated cellular activity in colorectal cancer models. It is supplied as a light yellow to yellow solid for laboratory research, provided with high purity and recommended cold storage for long-term stability.
  • Potent CDK2 and CDK5 inhibition with IC50s of ~6.0 nM and ~8.8 nM.
  • Demonstrated cellular activity in colorectal cancer cell lines.
  • High purity suitable for research applications.
  • Light yellow to yellow solid for ease of handling.
  • Molecular formula C19H19N3O and molecular weight 305.37.
  • Recommended cold storage for long-term stability.