Learn More
Medchemexpress LLC CDK5 inhibitor 20-223 | 865317-30-2 | MFCD32857141 | 99.6% | 305.37 | C19H19N3O | 50MG
SDP
Catalog No. 5000237619
Encompass_Preferred
CDK5 inhibitor 20-223 is a potent small-molecule inhibitor of cyclin-dependent kinases CDK2 and CDK5 (IC50 = 6.0 nM and 8.8 nM, respectively) with reported anti-colorectal cancer activity. It is supplied for research use in solid form or as a DMSO solution and is characterized by high purity and a defined molecular profile.
  • Potent CDK2 and CDK5 inhibition (IC50 6.0 nM and 8.8 nM).
  • Demonstrated anti-colorectal cancer activity in preclinical studies.
  • Available as solid powder and pre-dissolved DMSO solution.
  • High purity suitable for research applications (≈99.6%).
  • Defined molecular formula and molecular weight for reproducible results.
Catalog No. 50-002-37619 Supplier Medchemexpress LLC Supplier No. HY12377250MG
CDK5 inhibitor 20-223 is a potent small-molecule inhibitor of cyclin-dependent kinases CDK2 and CDK5 (IC50 = 6.0 nM and 8.8 nM, respectively) with reported anti-colorectal cancer activity. It is supplied for research use in solid form or as a DMSO solution and is characterized by high purity and a defined molecular profile.
  • Potent CDK2 and CDK5 inhibition (IC50 6.0 nM and 8.8 nM).
  • Demonstrated anti-colorectal cancer activity in preclinical studies.
  • Available as solid powder and pre-dissolved DMSO solution.
  • High purity suitable for research applications (≈99.6%).
  • Defined molecular formula and molecular weight for reproducible results.