Learn More
Carnabio Usa Inc CDK5/P25/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0410610
Encompass
CDK5/p25, Full-length, Wild type, Amino Acid 1-292, NP_004926.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0721