Learn More
Eurofins Discoverx Cdk5/p25, active, 250 µg. (14-516M)
Supplier:  Eurofins Discoverx 14516M
Encompass
Complex of N-terminal 6xHis-tagged recombinant, human full length cdk5 and N-terminal GST-tagged recombinant, human full length p25. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8902