Learn More
Enzo Life Sciences CDK5 substrate (biotinylated) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2640001
Encompass_Preferred
This peptide, Biotin-Ahx-PKTPKKAKKL, is derived from histone H1 and is a substrate for CDK5. The biotin allows the peptide to be used in kinase assays with streptavidine-bound membranes. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1321