Learn More
CPC Scientific Biotin-Pro-Lys-Thr-Pro-Lys-Lys-Ala-Lys-Lys-Leu-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific HIST082A
Encompass
The native peptide PKTPKKAKKL is derived from histone H1 peptide sequence that is docked in the active site of cyclin-dependent kinase 5. It is an effective CDK5 substrate (Km = 40 ?M). ONE-LETTER SEQUENCE: Biotin-PKTPKKAKKLMOLECULAR FORMULA: C63H113N17O14S1MOLECULAR WEIGHT:1364.75STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Lew, J. Biochem. 42, 849 (2003); Sharma, P. et al. J. Biol. Chem. 274, 9600 (1999)
Catalog No. 50-210-9526