Learn More
CPC Scientific H-Pro-Lys-Thr-Pro-Lys-Lys-Ala-Lys-Lys-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST081A
Encompass
The native peptide PKTPKKAKKL is derived from the histone H1 peptide sequence that is docked in the active site of cyclin-dependent kinase 5. It is an effective CDK5 substrate (Km = 40 M).SEQUENCE: H-Pro-Lys-Thr-Pro-Lys-Lys-Ala-Lys-Lys-Leu-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: PKTPKKAKKLMOLECULAR FORMULA: C53H99N15O12MOLECULAR WEIGHT: 1138.45STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [164669-07-2]SYNONYMS: RESEARCH AREA: MiscellaneousREFERENCES:Lew, J. Biochem. 42, 849 (2003)Sharma, P. et al. J. Biol. Chem. 274, 9600 (1999)SKU(s): HIST-081A, HIST-081B
Catalog No. 50-210-9524