Learn More
Cayman Chemical Cdk5 Substrate trifluoroacet
SDP
Catalog No. 502967537
Encompass_Preferred
A synthetic peptide (PKTPKKAKKL) corresponding to a sequence of histone H1; phosphorylated by Cdk5 with a Km value of 5 µM
Catalog No. 50-296-7537 Supplier Cayman Chemical Supplier No. 17477500 UG
A synthetic peptide (PKTPKKAKKL) corresponding to a sequence of histone H1; phosphorylated by Cdk5 with a Km value of 5 µM