Learn More
Cayman Chemical Cdk5 SubstratetrIfluoROCt 10mg
SDP
Catalog No. 502967208
Encompass_Preferred
A synthetic peptide (PKTPKKAKKL) corresponding to a sequence of histone H1; phosphorylated by Cdk5 with a Km value of 5 µM
Catalog No. 50-296-7208 Supplier Cayman Chemical Supplier No. 1747710 MG
A synthetic peptide (PKTPKKAKKL) corresponding to a sequence of histone H1; phosphorylated by Cdk5 with a Km value of 5 µM