Learn More
Sino Biological Recombinant Human CDKN2A/p16INK4a Protein (Full Length, GST Tag)
Catalog No. 502575596
Encompass_Preferred
Recombinant full length human p16INK4A was expressed in E. coli cells using a N-terminal GST tag.
Catalog No. 50-257-5596 Supplier Sino Biological Supplier No. C3030G1000
Add to Cart
Add to Cart
Request a Quote
Recombinant full length human p16INK4A was expressed in E. coli cells using a N-terminal GST tag.