Learn More
Aviva Systems Biology Corp CDK6 Antibody - C-terminal region (AVARP03005_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03005_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CDK6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CDK6. Alternate names: MGC59692, PLSTIREApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-62504