Learn More
Biomatik Corporation Cdk6 Recombinant Antibody
Supplier:  Biomatik Corporation RAC07062
Encompass_Preferred
Cdk6 Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to CDK6. The Cdk6 Recombinant Antibody has been validated for the following applications: ELISA, WB, IP. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: A Synthesized Peptide Derived From Human Cdk6. Applications: ELISA, WB, IP. Target Synonyms: CDK6. Accession Number: Q00534. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6845