Learn More
Carnabio Usa Inc CDK6/CYCD3/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0410710
Encompass
CDK6/CycD3, Full-length, Wild type, Amino Acid 1-326, NP_001250.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0722