Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CDK6/Cyclin D1, GST Tag Recombinant Protein

Catalog No. PR8422B
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR8422B 100 μg
PV4401 10 μg
PV4437 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR8422B Supplier Invitrogen™ Supplier No. PR8422B
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

Cyclin-dependent kinase 6 (CDK6) and Cyclin D1 are critical components of the cell cycle regulation, particularly in the transition from the G1 to the S phase. CDK6 is a protein kinase that requires association with cyclins, such as Cyclin D1, to become enzymatically active. This partnership is essential for phosphorylating target substrates that drive cell cycle progression. CDK6 and Cyclin D1 together form complexes that regulate the retinoblastoma protein (Rb) pathway, which is a key mechanism controlling cell proliferation. The expression and activation of CDK6/Cyclin D1 complexes are tightly regulated, and imbalances can contribute to oncogenesis. Structural studies have shown that CDK6 has a unique C-terminal region crucial for its nuclear transport and localization, which is essential for its function. Cyclin D1's dynamics-its expression, accumulation, and degradation-are pivotal in its regulatory roles, especially its interactions with CDK4 and CDK6. In cancer, genetic alterations affecting CDK6 and Cyclin D1 can lead to uncontrolled cell division, emphasizing their significance in cancer progression and as potential therapeutic targets.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P24385, Q00534
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 150mM NaCl, 0.5mM EDTA, 2mM DTT, 0.02% Triton X-100, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 1021, 595
Molecular Weight (g/mol) 66.4-61.2 kDa
Name Human CDK6/Cyclin D1, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 5830411I20; AI504062; CDK6; CDKN6; Cell division protein kinase 6; CR2 protein kinase; Crk2; cyclin dependent kinase 6; cyclin-dependent kinase 6; MCPH12; MGC59692; PLSTIRE; Serine/threonine-protein kinase PLSTIRE
Common Name CDK6/Cyclin D1
Gene Symbol CCND1, CDK6
Kinase Family CDK and CHK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: CMCG Group
Shipping Condition Dry Ice
Target Entry CDK6/Cyclin D1
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Research Category Cell Cycle & Proliferation, Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.