Learn More
Eurofins Discoverx Cdk6/Cyclin D3, active, 10 µg. (14-519)
Supplier:  Eurofins Discoverx 14519
Encompass
N-terminal 6xHis-tagged recombinant, human full length cdk6 complexed with N-terminal GST-tagged full length human cyclin D3. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8907