Learn More
Sino Biological Recombinant Human CDK4 & Cyclin D3 Protein (Full Length, GST Tag), Active
Catalog No. 502575587
Encompass_Preferred
Recombinant full length human CDK4 and CyclinD3 were co-expressed by baculovirus in Sf9 cells using an N-terminal GST tag on both proteins.
Catalog No. 50-257-5587 Supplier Sino Biological Supplier No. C3118G50
Recombinant full length human CDK4 and CyclinD3 were co-expressed by baculovirus in Sf9 cells using an N-terminal GST tag on both proteins.