Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CDK7/9 tide | 99.9% | 2690.00 | 5 MG
SDP

Catalog No. 5000424176
Encompass_Preferred

CDK7/9 tide is a peptide substrate designed for use with CDK7 or CDK9. It features a molecular weight of 2690.00 and appears as a white to off-white solid. The peptide's sequence shortening is YSPTSPSYSPTSPSYSPTSPSKKKK. It is soluble in H2O at 10 mg/mL (3.72 mM), requiring ultrasonic assistance for dissolution.

  • Peptide substrate for CDK7 or CDK9
  • Molecular weight: 2690.00
  • Appearance: white to off-white solid
  • Sequence shortening: YSPTSPSYSPTSPSYSPTSPSKKKK
  • Soluble in H2O: 10 mg/mL (3.72 mM)
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-24176 Supplier Medchemexpress LLC Supplier No. HYP25595MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CDK7/9 tide is a peptide substrate designed for use with CDK7 or CDK9. It features a molecular weight of 2690.00 and appears as a white to off-white solid. The peptide's sequence shortening is YSPTSPSYSPTSPSYSPTSPSKKKK. It is soluble in H2O at 10 mg/mL (3.72 mM), requiring ultrasonic assistance for dissolution.

  • Peptide substrate for CDK7 or CDK9
  • Molecular weight: 2690.00
  • Appearance: white to off-white solid
  • Sequence shortening: YSPTSPSYSPTSPSYSPTSPSKKKK
  • Soluble in H2O: 10 mg/mL (3.72 mM)
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.