Learn More
Aviva Systems Biology Corp CDK7 Antibody - C-terminal region (AVARP03009_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03009_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CDK7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CDK7. Alternate names: CAK1, CDKN7, MO15, STK1, p39MO15, HCAKApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-53992