Learn More
Eurofins Discoverx Cdk7/Cyclin H/MAT1 (CAK complex), active, 10 µg. (14-476)
Supplier:  Eurofins Discoverx 14476
Encompass
Complex of recombinant, human full length cdk7, containing a C-terminal 6xHis-tag, recombinant, human full length cyclin H untagged and recombinant, human full length MAT1 containing an N-terminal GST-tag. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8843