Learn More
Medchemexpress LLC Cdk7-in-2 | 2326428-19-5 | 98.9% | 497.63 g/mol | C26H39N7O3 | 5 MG
SDP
Catalog No. 5000221187
Encompass_Preferred
CDK7-IN-2 is a selective small-molecule inhibitor of cyclin-dependent kinase 7 (CDK7) used for biochemical and cellular research. It exhibits potent CDK7 inhibitory activity and antiproliferative effects in cancer cell models, and is supplied for research use only.
  • Selective CDK7 inhibitor suitable for biochemical and cellular assays.
  • High reported purity (~98.9%) enabling reproducible results.
  • Soluble in DMSO for convenient stock solution preparation.
  • Suitable for in vitro cancer biology and transcriptional regulation studies.
  • Provided in small research-friendly quantities for assay development.
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-002-21187 Supplier Medchemexpress LLC Supplier No. HY1435875MG
CDK7-IN-2 is a selective small-molecule inhibitor of cyclin-dependent kinase 7 (CDK7) used for biochemical and cellular research. It exhibits potent CDK7 inhibitory activity and antiproliferative effects in cancer cell models, and is supplied for research use only.
  • Selective CDK7 inhibitor suitable for biochemical and cellular assays.
  • High reported purity (~98.9%) enabling reproducible results.
  • Soluble in DMSO for convenient stock solution preparation.
  • Suitable for in vitro cancer biology and transcriptional regulation studies.
  • Provided in small research-friendly quantities for assay development.
  • Intended for research use only; not for clinical or human use.