Learn More
Medchemexpress LLC Cdk7-in-21 | 2766124-39-2 | C33H36FN9O2 | 1 MG
SDP
Catalog No. 5000373759
Encompass_Preferred
Q901 (CDK7-IN-21) is a selective and potent CDK7 inhibitor with an IC50 of 10 nM. It disrupts MYC and E2F-driven cancer, such as colon cancer and lung cancer, by inhibiting CDK7.
  • Selective and potent CDK7 inhibitor (IC50 of 10 nM)
  • Disrupts MYC and E2F-driven cancer progression
  • Inhibits proliferation of MCF-7 breast cancer cells
  • Induces DNA damage accumulation in MCF-7 cells
  • Synergistically inhibits proliferation of various cancer cells when combined with other therapies
  • Inhibits tumor growth in HCT116 colon cancer xenograft models
  • Enhances tumor growth inhibition in combination with relevant drugs
Catalog No. 50-003-73759 Supplier Medchemexpress LLC Supplier No. HY1532781MG
Q901 (CDK7-IN-21) is a selective and potent CDK7 inhibitor with an IC50 of 10 nM. It disrupts MYC and E2F-driven cancer, such as colon cancer and lung cancer, by inhibiting CDK7.
  • Selective and potent CDK7 inhibitor (IC50 of 10 nM)
  • Disrupts MYC and E2F-driven cancer progression
  • Inhibits proliferation of MCF-7 breast cancer cells
  • Induces DNA damage accumulation in MCF-7 cells
  • Synergistically inhibits proliferation of various cancer cells when combined with other therapies
  • Inhibits tumor growth in HCT116 colon cancer xenograft models
  • Enhances tumor growth inhibition in combination with relevant drugs