Learn More
Cayman Chemical Cdk7humnrecombInt 100ug
SDP
Catalog No. 502945302
Encompass_Preferred
Source: Recombinant human N-terminal GST-tagged Cdk7 expressed in insect cells • Amino acids: 1-346 (full length) • MW: 65.6 kDa
Catalog No. 50-294-5302 Supplier Cayman Chemical Supplier No. 42000100 UG
Source: Recombinant human N-terminal GST-tagged Cdk7 expressed in insect cells • Amino acids: 1-346 (full length) • MW: 65.6 kDa