Learn More
Aviva Systems Biology Corp CDK8 Antibody - C-terminal region (AVARP03050_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03050_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CDK8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CDK8. Alternate names: K35ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Zebrafish
Catalog No. 50-014-62530