Learn More
Eurofins Discoverx Cdk9/Cyclin T1, active, 250 µg. (14-685M)
Supplier:  Eurofins Discoverx 14685M
Encompass
C-terminal 6xHis-tagged recombinant, human full length cdk9 co-expressed with untagged, full length human cyclin T1. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9186