Learn More
Medchemexpress LLC HY-13231 5mg Medchemexpress, CDK9-IN-1 CAS:1415559-43-1 Purity:>98%
SDP
Catalog No. 502028963
Encompass_Preferred
Medchemexpress, HY-13231 5mg CDK9-IN-1 CAS:1415559-43-1 CDK9-IN-1 is a novel, selective CDK9 inhibitor for the treatment of HIV infection, with an IC50 of 39 nM for CDK9/CycT1, extracted from reference, compound 87. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8963 Supplier Medchemexpress LLC Supplier No. HY132315MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-13231 5mg CDK9-IN-1 CAS:1415559-43-1 CDK9-IN-1 is a novel, selective CDK9 inhibitor for the treatment of HIV infection, with an IC50 of 39 nM for CDK9/CycT1, extracted from reference, compound 87. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.