Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CDK9-IN-13 | 2768712-71-4 | 99.9% | 461.60 | 50 MG
SDP

Catalog No. 5000352359
Encompass_Preferred

CDK9-IN-13 is a potent and selective CDK9 inhibitor, demonstrating an IC50 of <3 nM. It also shows inhibitory activity against CDK1, CDK2, CDK7, and CDK12. The compound exhibits short half-lives in rodents and is intended for research use only.

  • Potent and selective CDK9 inhibitor
  • Inhibits CDK1, CDK2, CDK7, and CDK12
  • Suitable for research use only
  • Short half-lives in rodents
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (2.5 mg/mL)

Catalog No. 50-003-52359 Supplier Medchemexpress LLC Supplier No. HY13998050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CDK9-IN-13 is a potent and selective CDK9 inhibitor, demonstrating an IC50 of <3 nM. It also shows inhibitory activity against CDK1, CDK2, CDK7, and CDK12. The compound exhibits short half-lives in rodents and is intended for research use only.

  • Potent and selective CDK9 inhibitor
  • Inhibits CDK1, CDK2, CDK7, and CDK12
  • Suitable for research use only
  • Short half-lives in rodents
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (2.5 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.