Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cdk9-in-13 | 2768712-71-4 | 99.94% | 461.60 | 100 MG
SDP

Catalog No. 5000352093
Encompass_Preferred

CDK9-IN-13 (compound 38) is a potent and selective CDK9 inhibitor, with an IC50 of <3 nM. It exhibits short half-lives in rodents. It is suitable for laboratory centrifugation and storage.

  • Potent and selective cdk9 inhibitor
  • Ic50 of <3 nm
  • Short half-lives in rodents
  • Cdk1: 0.14 μm
  • Cdk2: 0.72 μm
  • Cdk7: 0.40 μm
  • Cdk12: 0.055 μm

Catalog No. 50-003-52093 Supplier Medchemexpress LLC Supplier No. HY139980100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CDK9-IN-13 (compound 38) is a potent and selective CDK9 inhibitor, with an IC50 of <3 nM. It exhibits short half-lives in rodents. It is suitable for laboratory centrifugation and storage.

  • Potent and selective cdk9 inhibitor
  • Ic50 of <3 nm
  • Short half-lives in rodents
  • Cdk1: 0.14 μm
  • Cdk2: 0.72 μm
  • Cdk7: 0.40 μm
  • Cdk12: 0.055 μm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.