Learn More
Medchemexpress LLC Cdk9-in-13 (compound 38) | 2768712-71-4 | MFCD11162707 | 99.9% | C27H35N5O2 | 10MG
SDP
Catalog No. 5000258457
Encompass_Preferred
CDK9-IN-13 is a research small-molecule inhibitor selective for cyclin-dependent kinase 9 (CDK9). Reported as compound 38, it inhibits CDK9 with an IC50 of <3 nM and has been characterized in preclinical rodent studies where it exhibits short in vivo half-lives. Intended for in vitro and early preclinical research use only.
  • Potent CDK9 inhibition with IC50 <3 nM.
  • Selective activity toward CDK9 for target validation studies.
  • High supplied purity (≈99.94%), limiting impurity-related variability.
  • Defined molecular properties (C27H35N5O2; MW 461.60 g·mol⁻1).
  • Offered in multiple small-scale quantities suitable for preclinical experiments.
Catalog No. 50-002-58457 Supplier Medchemexpress LLC Supplier No. HY13998010MG
CDK9-IN-13 is a research small-molecule inhibitor selective for cyclin-dependent kinase 9 (CDK9). Reported as compound 38, it inhibits CDK9 with an IC50 of <3 nM and has been characterized in preclinical rodent studies where it exhibits short in vivo half-lives. Intended for in vitro and early preclinical research use only.
  • Potent CDK9 inhibition with IC50 <3 nM.
  • Selective activity toward CDK9 for target validation studies.
  • High supplied purity (≈99.94%), limiting impurity-related variability.
  • Defined molecular properties (C27H35N5O2; MW 461.60 g·mol⁻1).
  • Offered in multiple small-scale quantities suitable for preclinical experiments.