Learn More
TARGETMOL CHEMICALS INC CDK9-IN-2 5MG
SDP
Catalog No. 502581271
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CDK9-IN-2 is a special CDK9 inhibitor extracted from patent WO/2012131594A1 (compound CDKI(8)) and has an IC50 of 5 nM and 7 nM in A2058 skin cell line (72 hours) and H929 multiple myeloma cell line (72 hours) respectively. purity: 99%
Catalog No. 50-258-1271 Supplier TARGETMOL CHEMICALS INC Supplier No. T149185MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CDK9-IN-2 is a special CDK9 inhibitor extracted from patent WO/2012131594A1 (compound CDKI(8)) and has an IC50 of 5 nM and 7 nM in A2058 skin cell line (72 hours) and H929 multiple myeloma cell line (72 hours) respectively. purity: 99%