Learn More
Medchemexpress LLC Cdk9-in-7 | 2369981-71-3 | 98.1% | 547.71 | C29H37N7O2S | 5MG
SDP
Catalog No. 5000234938
Encompass_Preferred
CDK9-IN-7 is a selective, potent, orally active small-molecule inhibitor of CDK9/cyclin T for research use. It exhibits strong biochemical potency (IC50 = 11 nM), preferential selectivity versus other CDKs, and has demonstrated antitumor activity in preclinical non-small-cell lung cancer models by inducing apoptosis, causing G2 cell-cycle arrest, and reducing cancer stemness.
  • Selective CDK9/cyclin T inhibition with reduced activity against CDK4 and CDK6
  • Potent biochemical activity (IC50 = 11 nM) for target engagement studies
  • Demonstrated antitumor effects in NSCLC models, including apoptosis and G2 arrest
  • Suitable for in vitro and in vivo research, including oral administration studies
  • Available as powder and as a 10 mM solution in DMSO for screening
  • High purity (~98.1%) for reliable experimental results
Catalog No. 50-002-34938 Supplier Medchemexpress LLC Supplier No. HY1262515MG
CDK9-IN-7 is a selective, potent, orally active small-molecule inhibitor of CDK9/cyclin T for research use. It exhibits strong biochemical potency (IC50 = 11 nM), preferential selectivity versus other CDKs, and has demonstrated antitumor activity in preclinical non-small-cell lung cancer models by inducing apoptosis, causing G2 cell-cycle arrest, and reducing cancer stemness.
  • Selective CDK9/cyclin T inhibition with reduced activity against CDK4 and CDK6
  • Potent biochemical activity (IC50 = 11 nM) for target engagement studies
  • Demonstrated antitumor effects in NSCLC models, including apoptosis and G2 arrest
  • Suitable for in vitro and in vivo research, including oral administration studies
  • Available as powder and as a 10 mM solution in DMSO for screening
  • High purity (~98.1%) for reliable experimental results