Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cdk9-in-7 | 2369981-71-3 | 98.1% | 547.71 g·mol⁻¹ | C29H37N7O2S | 10MG
SDP

Catalog No. 5000234969
Encompass_Preferred

CDK9-IN-7 is a selective, potent small-molecule inhibitor of cyclin-dependent kinase 9 (CDK9) intended for research use. It exhibits nanomolar potency (IC50 ≈ 11 nM) and has been reported to induce apoptosis and cell-cycle arrest in cancer cell models. Supplied as a powder, it is suitable for preclinical assays and mechanistic studies.

  • Selective CDK9 inhibition with reported IC50 ≈ 11 nM.
  • High purity approximately 98.1% suitable for research assays.
  • Molecular formula C29H37N7O2S and molecular weight 547.71 g·mol⁻¹.
  • Provided as a powder form, typically available in 10 mg quantity.
  • Recommended storage: powder -20°C (long term) or 4°C (short term).
  • Common applications include cell cycle, apoptosis, and transcriptional regulation studies.

Catalog No. 50-002-34969 Supplier Medchemexpress LLC Supplier No. HY12625110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CDK9-IN-7 is a selective, potent small-molecule inhibitor of cyclin-dependent kinase 9 (CDK9) intended for research use. It exhibits nanomolar potency (IC50 ≈ 11 nM) and has been reported to induce apoptosis and cell-cycle arrest in cancer cell models. Supplied as a powder, it is suitable for preclinical assays and mechanistic studies.

  • Selective CDK9 inhibition with reported IC50 ≈ 11 nM.
  • High purity approximately 98.1% suitable for research assays.
  • Molecular formula C29H37N7O2S and molecular weight 547.71 g·mol⁻¹.
  • Provided as a powder form, typically available in 10 mg quantity.
  • Recommended storage: powder -20°C (long term) or 4°C (short term).
  • Common applications include cell cycle, apoptosis, and transcriptional regulation studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.