Learn More
TARGETMOL CHEMICALS INC CDK9-IN-8 5MG
SDP
Catalog No. 502581674
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CDK9-IN-8 is a highly potent and selective CDK9 inhibitor (IC50 12 nM). purity: 98%
Catalog No. 50-258-1674 Supplier TARGETMOL CHEMICALS INC Supplier No. T107465MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CDK9-IN-8 is a highly potent and selective CDK9 inhibitor (IC50 12 nM). purity: 98%