Learn More
Cayman Chemical Cdk9 InhIbItr 25mg
SDP
Catalog No. 502978446
Encompass_Preferred
A Cdk9 inhibitor (IC50 = 39 nM); selective for Cdk9 over Cdk1-7 (IC50s = >1,000 nM for all); inhibits PRRSV replication in PRRSV-infected primary porcine alveolar macrophages at 6 to 200 nM
Catalog No. 50-297-8446 Supplier Cayman Chemical Supplier No. 4251225 MG
Add to Cart
Add to Cart
A Cdk9 inhibitor (IC50 = 39 nM); selective for Cdk9 over Cdk1-7 (IC50s = >1,000 nM for all); inhibits PRRSV replication in PRRSV-infected primary porcine alveolar macrophages at 6 to 200 nM