Learn More
Cayman Chemical CDKCRK InhIbItr 5mg
SDP
Catalog No. 502983724
Encompass_Preferred
A CDK/CRK-specific kinase inhibitor (IC50s = 9-839 nM, in vitro); selective, exhibiting less than 20% inhibition of 60 non-CDK/CRK kinases, at a concentration of 1 μM; induces cell cycle arrest in the G1 phase, endoreduplication, and apoptosis in HCT116 cells; exhibits broad anti-tumor activity with an average GI50 value of 50 = 40 nM)
Catalog No. 50-298-3724 Supplier Cayman Chemical Supplier No. 211955 MG
Add to Cart
Add to Cart
A CDK/CRK-specific kinase inhibitor (IC50s = 9-839 nM, in vitro); selective, exhibiting less than 20% inhibition of 60 non-CDK/CRK kinases, at a concentration of 1 μM; induces cell cycle arrest in the G1 phase, endoreduplication, and apoptosis in HCT116 cells; exhibits broad anti-tumor activity with an average GI50 value of 50 = 40 nM)