Learn More
Selleck Chemical LLC CDKI-73
SDP
Catalog No. 5000002883
Encompass_Preferred
CDKI-73 (LS-007) is a potent CDK inhibitor in vitro with IC50 of 8 17 nM 3 27 nM 8 18 nM and 5 78 nM for CDK1 CDK2 CDK4 and CDK9 respectively CDKI-73 induces apoptosis in cancer cells CDKI-73 is an orally bioavailable and highly efficacious CDK9 inhibitor against acute myeloid leukemia
Catalog No. 50-000-02883 Supplier Selleck Chemical LLC Supplier No. S77735MG
CDKI-73 (LS-007) is a potent CDK inhibitor in vitro with IC50 of 8 17 nM 3 27 nM 8 18 nM and 5 78 nM for CDK1 CDK2 CDK4 and CDK9 respectively CDKI-73 induces apoptosis in cancer cells CDKI-73 is an orally bioavailable and highly efficacious CDK9 inhibitor against acute myeloid leukemia